Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242837.1 | NP_001229766.1 | 2 | 326 | ||
NM_012305.3 | NP_036437.1 | 2 | 326 | |||
XM_011519929.1 | XP_011518231.1 | 2 | 83 | |||
XM_011519930.2 | XP_011518232.1 | 2 | 287 | |||
GenBank mRNA | AB020706.2 | - | 2 | 113 | ||
AK001986.1 | - | 2 | 255 | |||
AK027891.1 | - | 2 | 235 | |||
AK223558.1 | - | 2 | 254 | |||
AK293763.1 | - | 2 | 185 | |||
AK299364.1 | - | 2 | 104 | |||
BC006155.1 | - | 2 | 254 | 3627239 | ||
BG705558.1 | - | 2 | 294 | |||
BI462785.1 | - | 2 | 304 | |||
DB461570.1 | - | 2 | 326 |