Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144730.1 | NP_653331.1 | 7 | 3265 | ||
XM_006252189.2 | XP_006252251.1 | 6 | 2845 | |||
GenBank mRNA | L22761.1 | - | 7 | 3265 |