Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003466.3 | NP_003457.1 | 4 | 435 | ||
NM_013952.3 | NP_039246.1 | 4 | 435 | |||
NM_013953.3 | NP_039247.1 | 4 | 435 | |||
NM_013992.3 | NP_054698.1 | 4 | 435 | |||
XM_011511790.1 | XP_011510092.1 | 4 | 467 | |||
XM_011511791.1 | XP_011510093.1 | 4 | 467 | |||
XM_011511792.1 | XP_011510094.1 | 4 | 467 | |||
XM_011511793.1 | XP_011510095.1 | 4 | 467 | |||
XM_011511794.1 | XP_011510096.1 | 4 | 436 | |||
XM_017004878.1 | XP_016860367.1 | 4 | 467 | |||
XM_017004879.1 | XP_016860368.1 | 4 | 436 | |||
GenBank mRNA | AK225538.1 | - | 4 | 435 | ||
AK292191.1 | - | 4 | 432 | |||
AK309154.1 | - | 4 | 433 | |||
AU141429.1 | - | 4 | 435 | |||
BC001060.2 | - | 4 | 426 | 2963877 | ||
BG398792.1 | - | 437 | ||||
KC733810.1 | - | 4 | 532 | |||
KU178404.1 | - | 3 | 269 | |||
KU178405.1 | - | 3 | 269 | |||
L19606.1 | - | 3 | 279 | |||
S77904.1 | - | 4 | 429 | |||
S77905.1 | - | 4 | 429 | |||
S77906.2 | - | 4 | 429 | |||
X69699.1 | - | 4 | 429 |