Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177425.3 | NP_803174.2 | 4, 5 | 468 | ||
GenBank mRNA | BC086356.1 | - | 4, 5 | 482 | 7303714 | |
U44948.1 | - | 4, 5 | 473 |