Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183328.2 | NP_899157.1 | 2, 3 | 90 | ||
XM_008762822.1 | XP_008761044.1 | 3, 4 | 336 | |||
XM_008762823.1 | XP_008761045.1 | 3, 4 | 272 | |||
XM_008762824.1 | XP_008761046.1 | 3, 4 | 216 | |||
XM_008762825.1 | XP_008761047.1 | 3, 4 | 204 | |||
XM_008762826.1 | XP_008761048.1 | 3, 4 | 155 | |||
XM_008762827.1 | XP_008761049.1 | 3, 4 | 336 | |||
XM_008762828.1 | XP_008761050.1 | 3, 4 | 211 | |||
XM_008762829.1 | XP_008761051.1 | 2, 3 | 135 | |||
GenBank mRNA | M94557.1 | - | 2, 3 | 144 |