Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012310.4 | NP_036442.3 | 3 | 334 | ||
GenBank mRNA | AB208797.1 | - | 3 | 234 | ||
AF071592.2 | - | 3 | 251 | |||
AF179308.1 | - | 2 | 191 | |||
AJ271784.1 | - | 2 | 197 | |||
AK313133.1 | - | 2 | 567 | |||
BC003664.2 | - | 3 | 272 | |||
BC038459.1 | - | 3 | 271 | |||
BC049218.1 | - | 3 | 240 | |||
BC050548.1 | - | 3 | 233 | 4538604 | ||
DA021880.1 | - | 3 | 318 |