Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012490.2 | NP_036622.2 | 2 | 143 | ||
XM_006242178.2 | XP_006242240.1 | 2 | 1025 | |||
XM_008765665.1 | XP_008763887.1 | 3 | 1340 | |||
XM_008765666.1 | XP_008763888.1 | 3 | 865 | |||
GenBank mRNA | BC097345.1 | - | 2 | 143 | 7455117 | |
X59254.1 | - | 2 | 144 |