Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012492.2 | NP_036624.2 | 1 | 635 | ||
GenBank mRNA | AY057895.1 | - | 883 | |||
AY057896.1 | - | 1 | 884 | |||
BC086538.1 | - | 1 | 635 | 7114502 | ||
J03024.1 | - | 1 | 537 |
Set Membership: |
G-protein coupled receptor |