Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012493.2 | NP_036625.2 | 1 | 97 | ||
GenBank mRNA | BC097344.1 | - | 1 | 80 | 7459964 | |
DY309525.1 | - | 1 | 97 | |||
X02361.1 | - | 1 | 101 |