Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012496.2 | NP_036628.2 | 5 | 547 | ||
GenBank mRNA | BC081697.1 | - | 5 | 547 | 7109764 | |
FQ209505.1 | - | 5 | 567 | |||
FQ209538.1 | - | 5 | 567 | |||
FQ209583.1 | - | 5 | 567 | |||
FQ209594.1 | - | 5 | 567 | |||
FQ209615.1 | - | 5 | 567 | |||
FQ209621.1 | - | 5 | 565 | |||
FQ209639.1 | - | 5 | 567 | |||
FQ209643.1 | - | 5 | 564 | |||
FQ209782.1 | - | 5 | 567 | |||
FQ209819.1 | - | 5 | 577 | |||
FQ210002.1 | - | 5 | 567 | |||
FQ210849.1 | - | 5 | 567 | |||
FQ211083.1 | - | 5 | 567 | |||
FQ211156.1 | - | 5 | 567 | |||
FQ211159.1 | - | 5 | 567 | |||
FQ211293.1 | - | 3 | 245 | |||
FQ218139.1 | - | 5 | 567 | |||
FQ218217.1 | - | 5 | 567 | |||
FQ218348.1 | - | 5 | 573 | |||
FQ218402.1 | - | 5 | 569 | |||
FQ218522.1 | - | 5 | 567 | |||
FQ218551.1 | - | 5 | 567 | |||
FQ218592.1 | - | 1 | 65 | |||
FQ218665.1 | - | 5 | 567 | |||
FQ218885.1 | - | 5 | 567 | |||
FQ218911.1 | - | 5 | 567 | |||
FQ219077.1 | - | 3 | 251 | |||
FQ219096.1 | - | 5 | 567 | |||
FQ219116.1 | - | 5 | 567 | |||
FQ219236.1 | - | 5 | 566 | |||
FQ219453.1 | - | 5 | 567 | |||
FQ219555.1 | - | 5 | 567 | |||
FQ219574.1 | - | 5 | 567 | |||
FQ219585.1 | - | 5 | 567 | |||
FQ219640.1 | - | 5 | 565 | |||
FQ219646.1 | - | 5 | 567 | |||
FQ219703.1 | - | 5 | 566 | |||
M10149.1 | - | 5 | 558 |