Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012499.1 | NP_036631.1 | 1 | 139 | ||
XM_006254508.2 | XP_006254570.1 | 2 | 186 | |||
XM_006254509.2 | XP_006254571.1 | 2 | 155 | |||
XM_006254511.2 | XP_006254573.1 | 2 | 186 | |||
XM_006254521.2 | XP_006254583.1 | 2 | 186 | |||
XM_008772015.1 | XP_008770237.1 | 1 | 33 | |||
GenBank mRNA | D38629.1 | - | 1 | 139 |