Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012319.3 | NP_036451.3 | 2 | 761 | ||
XM_011525900.1 | XP_011524202.1 | 2 | 1194 | |||
XM_011525901.1 | XP_011524203.1 | 3 | 1878 | |||
GenBank mRNA | AK304079.1 | - | 2 | 685 | ||
DC369482.1 | - | 1 | 246 | |||
U41060.2 | - | 3 | 590 |