Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012511.2 | NP_036643.2 | 2 | 964 | ||
XM_006253346.2 | XP_006253408.1 | 2 | 1640 | |||
GenBank mRNA | AB017790.1 | - | 2 | 964 | ||
AB017791.1 | - | 2 | 964 | |||
AB017792.1 | - | 2 | 964 | |||
AB017793.1 | - | 2 | 964 | |||
U08344.1 | - | 2 | 980 |