Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012320.3 | NP_036452.1 | 5 | 637 | ||
XM_011522979.2 | XP_011521281.1 | 6 | 706 | |||
GenBank mRNA | AB017494.1 | - | 5 | 586 | ||
AK001705.1 | - | 5 | 598 | |||
AK074828.1 | - | 5 | 637 | |||
AK127898.1 | - | 1 | 1037 | |||
AK222790.1 | - | 5 | 596 | |||
AL110209.1 | - | 5 | 618 | |||
AY358425.1 | - | 5 | 574 | |||
BC011640.2 | - | 2 | 151 | |||
BC062605.1 | - | 5 | 601 | 5162981 |