Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 3 | 379 | ||
XM_011530679.2 | XP_011528981.1 | 3 | 268 | |||
XM_011530680.2 | XP_011528982.1 | 3 | 268 | |||
XM_011530681.2 | XP_011528983.1 | 3 | 268 | |||
GenBank mRNA | AF136382.1 | - | 3 | 362 | ||
AF218778.1 | - | 1 | 164 | |||
AK293580.1 | - | 3 | 292 | |||
AK294265.1 | - | 3 | 269 | |||
BC009940.2 | - | 3 | 272 | 4100637 | ||
DB483256.1 | - | 3 | 381 |