Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB025186.1 | - | 3 | 353 | ||
AK302950.1 | - | 3 | 427 | |||
AK312342.1 | - | 3 | 373 | |||
BC011557.1 | - | 3 | 338 | 4561018 | ||
BM560331.1 | - | 3 | 407 | |||
CR536523.1 | - | 2 | 200 | |||
CR541845.1 | - | 2 | 200 | |||
HY011071.1 | - | 3 | 439 | |||
Y11174.1 | - | 3 | 224 |