Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012529.3 | NP_036661.3 | 340 | |||
GenBank mRNA | BC065307.1 | - | 322 | 5598839 | ||
BC070955.1 | - | 327 | 7104205 | |||
BC087656.1 | - | 339 | ||||
BC099814.1 | - | 324 | 7929525 | |||
BC127477.1 | - | 330 | 8367180 | |||
FM060701.1 | - | 345 | ||||
FQ213834.1 | - | 315 | ||||
FQ216091.1 | - | 1 | 59 | |||
M14400.1 | - | 352 | ||||
M57664.1 | - | 2 | 203 |