Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012556.2 | NP_036688.1 | 2 | 198 | ||
GenBank mRNA | BC086947.1 | - | 2 | 198 | 7325407 | |
FQ209859.1 | - | 2 | 200 | |||
FQ209983.1 | - | 2 | 207 | |||
FQ210522.1 | - | 2 | 207 | |||
FQ211310.1 | - | 2 | 207 | |||
FQ211342.1 | - | 2 | 168 | |||
M10951.1 | - | 1 | 61 | |||
M17899.1 | - | 2 | 166 | |||
M35991.1 | - | 2 | 199 | |||
V01235.1 | - | 2 | 199 |