Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270849.1 | NP_001257778.1 | 8, 9 | 1076 | ||
NM_001270850.1 | NP_001257779.1 | 7, 8 | 854 | |||
NM_012565.2 | NP_036697.1 | 7, 8 | 977 | |||
XM_006251179.2 | XP_006251241.1 | 7, 8 | 1822 | |||
GenBank mRNA | J04218.1 | - | 7, 8 | 936 | ||
KJ026953.1 | - | 7, 8 | 869 | |||
M25807.1 | - | 7, 8 | 854 | |||
X53588.1 | - | 7, 8 | 960 | |||
X53589.1 | - | 8, 9 | 1076 |