Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012566.1 | NP_036698.1 | 6 | 1007 | ||
XM_008769917.1 | XP_008768139.1 | 6 | 1248 | |||
XM_008769918.1 | XP_008768140.1 | 6 | 1083 | |||
XM_008769919.1 | XP_008768141.1 | 6 | 1965 | |||
XM_008769920.1 | XP_008768142.1 | 6 | 1965 | |||
GenBank mRNA | L06986.1 | - | 6 | 1007 |