Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109968.1 | NP_001103438.1 | 7, 8 | 1112 | ||
NM_012569.2 | NP_036701.2 | 7, 8 | 1112 | |||
XM_008767075.1 | XP_008765297.1 | 7, 8 | 1283 | |||
GenBank mRNA | AY083459.1 | - | 2, 3 | 167 | ||
M65150.1 | - | 7, 8 | 1119 |