Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012605.2 | NP_036737.1 | 6, 7 | 451 | ||
GenBank mRNA | FQ216891.1 | - | 6, 7 | 467 | ||
FQ216934.1 | - | 6, 7 | 467 | |||
FQ217090.1 | - | 6, 7 | 469 | |||
FQ217098.1 | - | 6, 7 | 467 | |||
FQ217115.1 | - | 4, 5 | 316 | |||
FQ217155.1 | - | 6, 7 | 467 | |||
FQ217219.1 | - | 6, 7 | 467 | |||
FQ217259.1 | - | 6, 7 | 467 | |||
FQ217349.1 | - | 5, 6 | 391 | |||
FQ217365.1 | - | 6, 7 | 466 | |||
FQ217368.1 | - | 6, 7 | 483 | |||
FQ217533.1 | - | 6, 7 | 495 | |||
FQ217703.1 | - | 6, 7 | 467 | |||
FQ217740.1 | - | 6, 7 | 483 | |||
FQ217876.1 | - | 1, 2 | 49 | |||
FQ217881.1 | - | 6, 7 | 464 | |||
FQ217944.1 | - | 6, 7 | 467 | |||
FQ218017.1 | - | 6, 7 | 467 | |||
FQ223762.1 | - | 6, 7 | 448 | |||
FQ223868.1 | - | 6, 7 | 467 | |||
FQ223911.1 | - | 6, 7 | 483 | |||
FQ223944.1 | - | 5, 6 | 333 | |||
FQ223946.1 | - | 6, 7 | 467 | |||
FQ223966.1 | - | 6, 7 | 467 | |||
FQ223985.1 | - | 6, 7 | 467 | |||
FQ224150.1 | - | 4, 5 | 310 | |||
FQ224231.1 | - | 6, 7 | 467 | |||
FQ224253.1 | - | 6, 7 | 466 | |||
FQ224391.1 | - | 6, 7 | 465 | |||
FQ224404.1 | - | 6, 7 | 465 | |||
FQ224511.1 | - | 6, 7 | 467 | |||
FQ224525.1 | - | 6, 7 | 486 | |||
FQ224781.1 | - | 6, 7 | 483 | |||
J00754.1 | - | 3, 4 | 203 |