Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012611.3 | NP_036743.3 | 2796 | |||
GenBank mRNA | AY211532.1 | - | 2607 | |||
D12520.1 | - | 2796 | ||||
D14051.1 | - | 2626 | ||||
D44591.1 | - | 2627 | ||||
D83661.1 | - | 2606 | ||||
L12562.1 | - | 2708 | ||||
U03699.1 | - | 2609 | ||||
U26686.1 | - | 2606 | ||||
X76881.1 | - | 2791 |
Gene Symbol: LOC100359515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768231.1 | XP_008766453.1 | 1607 |
Gene Symbol: LOC497963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246949.2 | XP_006247011.1 | 2661 |