Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012617.1 | NP_036749.1 | 1 | 314 | ||
GenBank mRNA | D16348.1 | - | 1 | 314 | ||
U00475.1 | - | 1 | 257 |
Set Membership: |
![]() |