Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012618.2 | NP_036750.1 | 2 | 144 | ||
XM_006232592.2 | XP_006232654.1 | 2 | 204 | |||
GenBank mRNA | FQ215053.1 | - | 2 | 144 | ||
FQ220722.1 | - | 2 | 144 | |||
FQ221062.1 | - | 2 | 144 | |||
FQ221355.1 | - | 2 | 144 | |||
FQ221406.1 | - | 2 | 145 | |||
FQ221428.1 | - | 2 | 144 | |||
FQ221508.1 | - | 2 | 144 | |||
FQ221699.1 | - | 2 | 147 | |||
FQ221706.1 | - | 2 | 145 | |||
FQ221757.1 | - | 2 | 145 | |||
FQ221860.1 | - | 2 | 145 | |||
FQ222027.1 | - | 2 | 145 | |||
FQ222069.1 | - | 2 | 144 | |||
FQ222186.1 | - | 2 | 144 | |||
FQ222219.1 | - | 2 | 145 | |||
FQ222585.1 | - | 2 | 144 | |||
FQ222875.1 | - | 2 | 144 | |||
FQ222962.1 | - | 2 | 145 | |||
FQ223009.1 | - | 2 | 144 | |||
FQ223047.1 | - | 2 | 148 | |||
FQ223108.1 | - | 2 | 127 | |||
FQ223128.1 | - | 2 | 145 | |||
FQ223234.1 | - | 2 | 144 | |||
FQ228364.1 | - | 2 | 146 | |||
FQ228506.1 | - | 2 | 144 | |||
FQ228607.1 | - | 2 | 146 | |||
FQ229007.1 | - | 2 | 143 | |||
FQ229874.1 | - | 2 | 144 | |||
J03628.1 | - | 2 | 145 | |||
X64022.1 | - | 1 | 87 |