Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271063.1 | NP_001257992.1 | 4 | 399 | ||
NM_001271064.1 | NP_001257993.1 | 3 | 273 | |||
NM_012621.4 | NP_036753.4 | 4 | 522 | |||
XM_006256790.2 | XP_006256852.1 | 3 | 2432 | |||
XM_006256791.2 | XP_006256853.1 | 4 | 607 | |||
XM_008773123.1 | XP_008771345.1 | 6 | 573 | |||
XM_008773124.1 | XP_008771346.1 | 6 | 569 | |||
XM_008773125.1 | XP_008771347.1 | 5 | 447 | |||
GenBank mRNA | AY707861.1 | - | 4 | 350 | ||
GQ422136.1 | - | 3 | 253 | |||
J04197.1 | - | 4 | 552 | |||
X15579.1 | - | 4 | 379 | |||
X15580.1 | - | 3 | 1377 | |||
Y00702.1 | - | 4 | 522 |