Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012623.2 | NP_036755.2 | 18 | 2274 | ||
XM_008762687.1 | XP_008760909.1 | 17 | 2412 | |||
XM_008762688.1 | XP_008760910.1 | 17 | 2411 | |||
XM_008762689.1 | XP_008760911.1 | 13 | 2051 | |||
XM_008762691.1 | XP_008760913.1 | 17 | 2407 | |||
GenBank mRNA | AY082609.1 | - | 18 | 2274 | ||
M81855.1 | - | 18 | 2344 |