Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076553.2 | NP_001070021.1 | 6 | 862 | ||
NM_001271158.1 | NP_001258087.1 | 5 | 697 | |||
NM_012627.3 | NP_036759.2 | 5 | 757 | |||
XM_006256511.2 | XP_006256573.1 | 6 | 1481 | |||
XM_006256512.2 | XP_006256574.1 | 7 | 984 | |||
XM_006256513.2 | XP_006256575.1 | 6 | 839 | |||
XM_006256515.2 | XP_006256577.1 | 6 | 1404 | |||
XM_008772976.1 | XP_008771198.1 | 7 | 1121 | |||
XM_008772977.1 | XP_008771199.1 | 7 | 1834 | |||
XM_008772978.1 | XP_008771200.1 | 6 | 1571 | |||
GenBank mRNA | AF413572.1 | - | 6 | 838 | ||
BC078880.1 | - | 5 | 697 | 7111554 | ||
FQ231722.1 | - | 5 | 753 | |||
M64092.1 | - | 5 | 708 |