Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012634.1 | NP_036766.1 | 1 | 452 | ||
XM_006256829.2 | XP_006256891.1 | 1 | 215 | |||
GenBank mRNA | M17259.1 | - | 1 | 193 | ||
M29393.1 | - | 1 | 193 | |||
X16555.1 | - | 1 | 452 |
Gene Symbol: LOC100910245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256848.2 | XP_006256910.2 | 1 | 302 | ||
XM_008773204.1 | XP_008771426.1 | 219 |