Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284277.1 | NP_001271206.1 | 3 | 285 | ||
NM_001284278.1 | NP_001271207.1 | 3 | 263 | |||
NM_012399.4 | NP_036531.1 | 3 | 285 | |||
XM_011530052.2 | XP_011528354.1 | 3 | 298 | |||
XM_017028707.1 | XP_016884196.1 | 3 | 258 | |||
GenBank mRNA | AK302367.1 | - | 3 | 249 | ||
AK313064.1 | - | 3 | 235 | |||
BC018704.1 | - | 3 | 209 | 4643509 | ||
BC031427.1 | - | 3 | 202 | 4341137 | ||
BU147254.1 | - | 3 | 246 | |||
CR456541.1 | - | 3 | 237 | |||
D30037.1 | - | 3 | 216 | |||
DC313909.1 | - | 3 | 263 | |||
HY176246.1 | - | 3 | 285 |