Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012654.1 | NP_036786.1 | 15, 16 | 2338 | ||
XM_008758677.1 | XP_008756899.1 | 15, 16 | 2426 | |||
GenBank mRNA | M85300.1 | - | 15, 16 | 2338 |