Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 26 | 4379 | ||
XM_005261909.1 | XP_005261966.1 | 27 | 4427 | |||
XM_005261910.2 | XP_005261967.1 | 27 | 4438 | |||
XM_005261911.4 | XP_005261968.1 | 26 | 4377 | |||
XM_006724413.2 | XP_006724476.1 | 28 | 4497 | |||
XM_011530682.2 | XP_011528984.1 | 27 | 4578 | |||
XM_017028703.1 | XP_016884192.1 | 28 | 4478 | |||
XM_017028704.1 | XP_016884193.1 | 27 | 4431 | |||
GenBank mRNA | AB002313.1 | - | 25 | 4296 | ||
AB075511.1 | - | 11 | 1489 | |||
AK025701.1 | - | 13 | 1747 | |||
AK056543.1 | - | 9 | 1548 | |||
AK074932.1 | - | 8 | 1015 | |||
AK126394.1 | - | 7 | 1088 | |||
BC004542.2 | - | 3 | 198 | |||
BC148225.1 | - | 4 | 558 | |||
BC148280.1 | - | 4 | 558 | |||
BT006887.1 | - | 3 | 182 | |||
S76730.1 | - | 4 | 531 |