Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012657.2 | NP_036789.2 | 5 | 1515 | ||
XM_006240458.2 | XP_006240520.1 | 5 | 1558 | |||
GenBank mRNA | BC062236.1 | - | 5 | 1519 | 6888893 | |
D00751.1 | - | 5 | 1468 | |||
FQ209380.1 | - | 5 | 1518 | |||
FQ209577.1 | - | 5 | 1517 | |||
FQ209663.1 | - | 4 | 1243 | |||
FQ218085.1 | - | 5 | 1520 | |||
FQ218647.1 | - | 4 | 1192 | |||
FQ218802.1 | - | 5 | 1517 | |||
FQ219296.1 | - | 5 | 1521 | |||
FQ219312.1 | - | 5 | 1519 | |||
FQ219600.1 | - | 5 | 1515 | |||
M14320.1 | - | 4 | 1013 | |||
M15916.1 | - | 5 | 1480 | |||
X05348.1 | - | 5 | 1502 | |||
X16358.1 | - | 5 | 1516 |