Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012665.1 | NP_036797.1 | 8 | 2273 | ||
GenBank mRNA | M64488.1 | - | 8 | 2273 |