Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114099.2 | NP_001107571.1 | 2 | 352 | ||
NM_001114100.2 | NP_001107572.1 | 2 | 352 | |||
NM_001243332.1 | NP_001230261.1 | 2 | 352 | |||
NM_012410.3 | NP_036542.1 | 2 | 352 | |||
NM_201575.3 | NP_963869.2 | 2 | 352 | |||
XM_005255252.2 | XP_005255309.1 | 2 | 352 | |||
XM_017023135.1 | XP_016878624.1 | 2 | 661 | |||
GenBank mRNA | AB105376.1 | - | 2 | 191 | ||
AJ245820.1 | - | 2 | 191 | |||
AJ245821.1 | - | 2 | 191 | |||
AJ245822.1 | - | 2 | 191 | |||
AK299103.1 | - | 2 | 349 | |||
AY358404.1 | - | 2 | 341 | |||
BC000567.2 | - | 2 | 152 | 3163764 | ||
DA188718.1 | - | 2 | 351 | |||
DA726021.1 | - | 2 | 349 | |||
EL733582.1 | - | 2 | 107 | |||
KJ534938.1 | - | 2 | 107 | |||
KJ534939.1 | - | 2 | 107 | |||
KJ534940.1 | - | 2 | 107 | |||
KJ534941.1 | - | 2 | 107 | |||
KJ534988.1 | - | 2 | 107 | |||
KJ534989.1 | - | 2 | 107 | |||
KJ535068.1 | - | 2 | 107 | |||
KJ535092.1 | - | 2 | 106 |