Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTR SIRNA Assays ›
Gene Symbol:
Ttr
Gene Name:
transthyretin
Gene Aliases:
Lr1, Tbpa
Chromosome Location:
Chr.18: 15532963 - 15542180 on Build Rnor_6.0
UniGene ID:
Rn.1404
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200193
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ttr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012681.2
NP_036813.2
4
532
XM_006254457.1
XP_006254519.1
5
589
GenBank mRNA
AF479660.1
-
4
467
BC086946.1
-
4
425
7322736
FQ209711.1
-
4
447
FQ209724.1
-
4
450
FQ209731.1
-
4
450
FQ209737.1
-
4
450
FQ209743.1
-
4
450
FQ209798.1
-
4
450
FQ209827.1
-
4
449
FQ209853.1
-
4
450
FQ209868.1
-
4
450
FQ209884.1
-
4
450
FQ209896.1
-
4
443
FQ209911.1
-
4
450
FQ209919.1
-
4
449
FQ209931.1
-
4
447
FQ209949.1
-
4
450
FQ209974.1
-
4
450
FQ209986.1
-
4
450
FQ209992.1
-
4
447
FQ210005.1
-
4
450
FQ210012.1
-
4
449
FQ210053.1
-
4
450
FQ210063.1
-
4
450
FQ210074.1
-
4
445
FQ210079.1
-
4
450
FQ210088.1
-
4
450
FQ210104.1
-
4
449
FQ210128.1
-
4
450
FQ210130.1
-
4
450
FQ210144.1
-
4
450
FQ210148.1
-
4
450
FQ210149.1
-
4
450
FQ210166.1
-
4
450
FQ210168.1
-
4
450
FQ210188.1
-
4
450
FQ210192.1
-
4
445
FQ210193.1
-
4
450
FQ210202.1
-
4
450
FQ210212.1
-
4
450
FQ210220.1
-
4
450
FQ210221.1
-
4
450
FQ210257.1
-
4
450
FQ210271.1
-
4
450
FQ210283.1
-
4
447
FQ210289.1
-
4
445
FQ210300.1
-
4
450
FQ210301.1
-
4
450
FQ210303.1
-
4
450
FQ210314.1
-
4
450
FQ210319.1
-
4
450
FQ210324.1
-
4
448
FQ210363.1
-
4
450
FQ210365.1
-
4
449
FQ210370.1
-
4
450
FQ210380.1
-
4
450
FQ210383.1
-
4
447
FQ210386.1
-
900
FQ210389.1
-
4
450
FQ210423.1
-
4
449
FQ210429.1
-
4
450
FQ210431.1
-
4
450
FQ210434.1
-
4
450
FQ210451.1
-
4
450
FQ210464.1
-
4
445
FQ210465.1
-
4
450
FQ210502.1
-
4
450
FQ210512.1
-
4
449
FQ210545.1
-
4
450
FQ210547.1
-
4
450
FQ210548.1
-
4
447
FQ210552.1
-
4
445
FQ210579.1
-
4
449
FQ210580.1
-
4
447
FQ210584.1
-
4
450
FQ210588.1
-
4
450
FQ210614.1
-
4
450
FQ210642.1
-
4
450
FQ210659.1
-
4
449
FQ210663.1
-
4
447
FQ210667.1
-
4
450
FQ210680.1
-
4
445
FQ210689.1
-
4
450
FQ210691.1
-
4
450
FQ210711.1
-
4
447
FQ210744.1
-
4
450
FQ210747.1
-
4
448
FQ210750.1
-
4
449
FQ210759.1
-
4
450
FQ210777.1
-
4
447
FQ210778.1
-
4
450
FQ210779.1
-
4
450
FQ210782.1
-
4
450
FQ210786.1
-
4
450
FQ210844.1
-
4
450
FQ210853.1
-
4
447
FQ210860.1
-
4
450
FQ210869.1
-
4
450
FQ210887.1
-
4
450
FQ210913.1
-
4
450
FQ210915.1
-
4
450
FQ210971.1
-
4
450
FQ210974.1
-
4
450
FQ210975.1
-
4
450
FQ210993.1
-
4
450
FQ211011.1
-
4
450
FQ211023.1
-
4
450
FQ211028.1
-
4
447
FQ211048.1
-
4
450
FQ211066.1
-
4
450
FQ211087.1
-
4
450
FQ211100.1
-
4
450
FQ211107.1
-
4
450
FQ211134.1
-
4
449
FQ211143.1
-
4
450
FQ211145.1
-
4
450
FQ211149.1
-
4
450
FQ211173.1
-
4
450
FQ211184.1
-
4
456
FQ211191.1
-
4
450
FQ211204.1
-
4
450
FQ211212.1
-
4
450
FQ211228.1
-
4
450
FQ211233.1
-
4
452
FQ211237.1
-
4
447
FQ211240.1
-
4
450
FQ211260.1
-
4
450
FQ211262.1
-
4
448
FQ211291.1
-
4
447
FQ211302.1
-
4
450
FQ211335.1
-
4
450
FQ211341.1
-
4
450
FQ211348.1
-
4
447
FQ211403.1
-
4
450
FQ211448.1
-
4
450
FQ211620.1
-
4
450
FQ212160.1
-
4
450
FQ212302.1
-
4
450
FQ213410.1
-
4
535
FQ217460.1
-
4
450
FQ218087.1
-
4
450
FQ218127.1
-
4
451
FQ218128.1
-
4
446
FQ218131.1
-
4
450
FQ218133.1
-
4
450
FQ218134.1
-
4
450
FQ218142.1
-
4
445
FQ218150.1
-
4
450
FQ218178.1
-
4
450
FQ218187.1
-
4
450
FQ218301.1
-
4
450
FQ218306.1
-
4
450
FQ218313.1
-
4
450
FQ218333.1
-
4
445
FQ218513.1
-
4
445
FQ218589.1
-
4
450
FQ218636.1
-
4
450
FQ218738.1
-
4
450
FQ218740.1
-
4
450
FQ218747.1
-
4
450
FQ218749.1
-
4
450
FQ218786.1
-
4
450
FQ218849.1
-
4
450
FQ218866.1
-
4
450
FQ218996.1
-
4
450
FQ219153.1
-
4
447
FQ219387.1
-
4
450
FQ219428.1
-
4
450
FQ223520.1
-
4
445
FQ223553.1
-
4
450
FQ224434.1
-
4
450
K03252.1
-
4
430
X14876.1
-
4
423
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
Gene Ontology Categories:
Function(s)
Process(es)
thyroid hormone metabolic process
retinol metabolic process
thyroid hormone transport
hormone activity
hormone binding
identical protein binding
protein heterodimerization activity
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-j25zq:80/100.66.77.222:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0