Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012683.2 | NP_036815.1 | 1 | 214 | ||
GenBank mRNA | AF461736.1 | - | 1 | 217 | ||
AY435128.1 | - | 1 | 198 | |||
S70360.1 | - | 1 | 234 |