Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012699.2 | NP_036831.2 | 2 | 353 | ||
XM_006240065.2 | XP_006240127.1 | 2 | 344 | |||
GenBank mRNA | BC070915.1 | - | 2 | 351 | 7104653 | |
FQ230361.1 | - | 1 | 106 | |||
X98993.2 | - | 2 | 353 |