Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012717.1 | NP_036849.1 | 11 | 1163 | ||
XM_006234438.2 | XP_006234500.1 | 11 | 1418 | |||
XM_006234439.2 | XP_006234501.1 | 10 | 1256 | |||
GenBank mRNA | L27487.1 | - | 11 | 1436 | ||
X70658.1 | - | 11 | 1163 |
Set Membership: |
G-protein coupled receptor |