Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Andpro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012718.2 | NP_036850.1 | 1 | 142 | ||
XM_006235184.2 | XP_006235246.1 | 1 | 381 | |||
GenBank mRNA | BC065311.1 | - | 1 | 122 | ||
M27901.1 | - | 1 | 126 | |||
M58167.1 | - | 1 | 143 |