Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012723.4 | NP_036855.3 | 539 | |||
GenBank mRNA | FQ104960.1 | - | 3 | 539 | ||
X15741.1 | - | 2 | 433 |