Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012737.1 | NP_036869.1 | 2 | 206 | ||
GenBank mRNA | BC091159.1 | - | 2 | 181 | 7373033 | |
FQ218620.1 | - | 2 | 207 | |||
M00002.1 | - | 2 | 206 |