Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199897.1 | NP_001186826.1 | 5 | 1262 | ||
NM_012261.3 | NP_036393.1 | 6 | 1394 | |||
GenBank mRNA | AK316114.1 | - | 5 | 1262 | ||
AL121740.1 | - | 6 | 1101 | |||
BC050727.1 | - | 6 | 1097 | 6044324 |