Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195071.1 | NP_001182000.1 | 9 | 1645 | ||
NM_001195072.1 | NP_001182001.1 | 10 | 1731 | |||
NM_012264.4 | NP_036396.2 | 9 | 1630 | |||
XM_011530112.2 | XP_011528414.2 | 10 | 1700 | |||
XM_011530113.1 | XP_011528415.1 | 9 | 1645 | |||
XM_011530114.1 | XP_011528416.1 | 9 | 1677 | |||
XM_011530115.2 | XP_011528417.1 | 8 | 1436 | |||
XM_011530117.1 | XP_011528419.1 | 10 | 1743 | |||
XM_011530118.1 | XP_011528420.1 | 10 | 1717 | |||
XM_011530119.1 | XP_011528421.1 | 8 | 1471 | |||
XM_017028754.1 | XP_016884243.1 | 9 | 1716 | |||
XM_017028755.1 | XP_016884244.1 | 9 | 1673 | |||
XM_017028756.1 | XP_016884245.1 | 10 | 1630 | |||
XM_017028757.1 | XP_016884246.1 | 9 | 1418 | |||
XM_017028759.1 | XP_016884248.1 | 8 | 1435 | |||
GenBank mRNA | AB097053.1 | - | 9 | 1601 | ||
AK075417.1 | - | 7 | 1118 | |||
AK095109.1 | - | 1 | 1950 | |||
AL096879.1 | - | 8 | 1323 | |||
AL117649.1 | - | 1 | 206 | |||
BC012983.2 | - | 5 | 954 | |||
BC015489.1 | - | 9 | 1586 | 3896682 | ||
BX648345.1 | - | 9 | 1681 | |||
CR627445.1 | - | 1 | 2832 | |||
CR749859.1 | - | 10 | 1637 |