Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012780.1 | NP_036912.1 | 12, 13 | 1186 | ||
XM_006232850.2 | XP_006232912.1 | 12, 13 | 1313 | |||
XM_006232852.2 | XP_006232914.1 | 12, 13 | 1403 | |||
XM_006232853.2 | XP_006232915.1 | 11, 12 | 1268 | |||
XM_008761280.1 | XP_008759502.1 | 12, 13 | 1313 | |||
XM_008761281.1 | XP_008759503.1 | 11, 12 | 1268 | |||
GenBank mRNA | AY264361.1 | - | 12, 13 | 1186 | ||
AY264362.1 | - | 11, 12 | 1141 | |||
AY264363.1 | - | 12, 13 | 1045 | |||
AY264364.1 | - | 12, 13 | 1171 | |||
AY264365.1 | - | 12, 13 | 1186 | |||
U08986.1 | - | 7, 8 | 893 | |||
U61184.1 | - | 12, 13 | 1186 |