Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040186.2 | NP_001035276.1 | 5 | 735 | ||
NM_012811.3 | NP_036943.1 | 4 | 567 | |||
GenBank mRNA | BC085754.1 | - | 4 | 520 | 7193796 | |
D84068.1 | - | 4 | 527 | |||
DQ455823.1 | - | 5 | 688 | |||
FQ134652.1 | - | 4 | 567 |