Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012816.2 | NP_036948.2 | 5 | 1440 | ||
GenBank mRNA | BC078885.1 | - | 5 | 1408 | 7110324 | |
BC091163.1 | - | 5 | 1409 | 7130642 | ||
BC127505.1 | - | 5 | 1429 | 7325347 | ||
U89905.1 | - | 5 | 1428 | |||
Y08172.1 | - | 5 | 1418 |