Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012289.3 | NP_036421.2 | 6 | 2162 | ||
NM_203500.1 | NP_987096.1 | 6 | 2191 | |||
XM_005260173.1 | XP_005260230.1 | 6 | 2330 | |||
XM_005260174.1 | XP_005260231.1 | 6 | 2345 | |||
XM_011528452.1 | XP_011526754.1 | 6 | 2147 | |||
GenBank mRNA | AF361886.1 | - | 6 | 2118 | ||
AK056204.1 | - | 6 | 2380 | |||
BC002417.2 | - | 5 | 2036 | 3163902 | ||
BC002930.1 | - | 6 | 2112 | 3955118 | ||
BC003156.1 | - | 4 | 867 | |||
BC015945.1 | - | 5 | 2047 | 3910857 | ||
BC021957.1 | - | 4 | 867 | |||
D50922.2 | - | 6 | 2118 |