Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017012.1 | NP_058708.1 | 5 | 1603 | ||
XM_006229659.2 | XP_006229721.1 | 5 | 1930 | |||
GenBank mRNA | D10891.1 | - | 5 | 1603 | ||
EU559292.1 | - | 4 | 1336 |
Set Membership: |
G-protein coupled receptor |