Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017017.2 | NP_058713.1 | 2 | 315 | ||
XM_006235979.2 | XP_006236041.1 | 2 | 410 | |||
GenBank mRNA | D90102.1 | - | 2 | 315 | ||
X54400.1 | - | 2 | 214 |